Order Entry
Italy
ContactUsLinkComponent
recombinant/native protein, ptk2 (human) recombinant protein, human ptk2 (np_722560.1, 1 a.a. - 527 a.a.) full-length recombinant pr otein with gst tag co-expressed with his-tagged wnk1 (np_061852.1, 1 a.a. - 491 a.a.) 1 * 5 µG
Catalog # ABNOP5710
Supplier:  Abnova
recombinant/native protein, ptk2 (human) recombinant protein, human ptk2 (np_722560.1, 1 a.a. - 527 a.a.) full-length recombinant pr otein with gst tag co-expressed with his-tagged wnk1 (np_061852.1, 1 a.a. - 491 a.a.) 1 * 5 µG
Catalog # ABNOP5710
Supplier:  Abnova