recombinant protein, pip5k3 (human) recombinant protein (p02), human pip5k3 full-length orf ( np_689884.1, 1 a.a. - 451 a.a.) recomb inant protein with gst-tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00200576-P02
Supplier: Abnova
Frequently Bought Together