Order Entry
Italy
Orders LinkContactUsLinkComponent
Polysorbate 20 NF, Multi-Compendial, J.T.Baker®

Polysorbate 20 NF, Multi-Compendial, J.T.Baker®

Glass bottle, amber, narrow mouth, 20 mL
Catalog # 4116-02
Supplier:   Avantor
Polysorbate 20 NF, Multi-Compendial, J.T.Baker®
Catalog # 4116-02
Supplier:  Avantor
CAS Number:  9005-64-5
Polysorbate 20 NF, Multi-Compendial, J.T.Baker®
Polysorbate 20 NF, Multi-Compendial, J.T.Baker®

Glass bottle, amber, narrow mouth, 20 mL

CAS Number:  9005-64-5
Price
159,35 €
Log in for availability
1 * 20 mL
Product Details & Documents
Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Density:1.11 g/cm³
  •  :Management of Change (MOC) category = R
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specification Test Results

  • GMP Manufactured Product:
    ≤2.0
  • Meets JPE Specifications:
    ≤0.25%
  • Meets N.F. Requirements:
    ≤10 ppm
  • Meets E.P. Chemical Specifications:
    ≤1 ppm
  • Meets B.P. Chemical Specifications:
    96 - 108
  • Store in airtight container, protected from light:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    40 - 50
  • NF - Acid Value:
    ≤ 2.0
  • NF - Residue on Ignition:
    ≤ 0.25 %
  • NF - Dioxane:
    ≤ 10 ppm
  • NF - Ethylene Oxide:
    ≤ 1 ppm
  • NF - Hydroxyl Value, mg KOH/g:
    96 - 108
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Saponification Value, mg KOH/g:
    40 - 50
  • NF - Caproic Acid (Composition of Fatty Acids):
    ≤ 1.0 %
  • NF - Caprylic Acid (Composition of Fatty Acids):
    ≤ 10.0 %
  • NF - Capric Acid (Composition of Fatty Acids):
    ≤ 10.0 %
  • NF - Lauric Acid (Composition of Fatty Acids):
    40.0 - 60.0 %
  • NF - Myristic Acid (Composition of Fatty Acids):
    14.0 - 25.0 %
  • NF - Palmitic Acid (Composition of Fatty Acids):
    7.0 - 15.0 %
  • NF - Stearic Acid (Composition of Fatty Acids):
    ≤ 11.0 %
  • NF - Oleic Acid (Composition of Fatty Acids):
    ≤ 11.0 %
  • NF - Linoleic Acid (Composition of Fatty Acids):
    ≤ 3.0 %
  • NF - Peroxide Value:
    ≤ 5.0
  • NF - Water (H₂O):
    ≤ 3.0 %
  • Endotoxin Concentration (EU/mL):
    ≤ 10
  • EP/BP - Acid Value:
    ≤ 2.0
  • EP/BP - Total Ash:
    ≤ 0.25 %
  • EP/BP - Composition of Fatty Acids - Caproic Acid:
    ≤ 1.0 %
  • EP/BP - Composition of Fatty Acids - Caprylic Acid:
    ≤ 10.0 %
  • EP/BP - Composition of Fatty Acids - Capric Acid:
    ≤ 10.0 %
  • EP/BP - Composition of Fatty Acids - Lauric Acid:
    40.0 - 60.0 %
  • EP/BP - Composition of Fatty Acids - Myristic Acid:
    14.0 - 25.0 %
  • EP/BP -Composition of Fatty Acids - Palmitic Acid:
    7.0 - 15.0 %
  • EP/BP - Composition of Fatty Acids - Stearic Acid:
    ≤ 7.0 %
  • EP/BP - Composition of Fatty Acids - Oleic Acid:
    ≤ 11.0 %
  • EP/BP - Composition of Fatty Acids - Linoleic Acid:
    ≤ 3.0 %
  • EP - Identification A:
    Passes Test
  • EP - Identification D:
    Passes Test
  • EP/BP - Hydroxyl Value:
    96 - 108
  • EP - Peroxide Value:
    ≤ 10.0
  • EP - Saponification Value:
    40 - 50
  • EP - Water (H₂O):
    ≤ 3.0 %
  • EP - Ethylene Oxide:
    ≤ 1 ppm
  • EP - Dioxan:
    ≤ 10 ppm
  • Appearance:
    Passes Test
  • pH of 5% Solution at 25°C:
    5.0 - 7.0
  • Arsenic (As):
    ≤ 1.000 ppm
  • Peroxide Value, meqO₂/kg:
    ≤ 2.0
  • Microbiological - Total Aerobic Microbial Count (cfu/g):
    ≤ 100
  • Microbiological - Escherichia Coli:
    Passes Test
  • Microbiological - Pseudomonas aeruginosa:
    Passes Test
  • Microbiological - Salmonella:
    Passes Test
  • Microbiological - Staphylococcus aureus:
    Passes Test
  • Microbiological - Total Yeast and Mold Count (cfu/g):
    ≤ 50
  • JPE - Identification 1:
    Passes Test
  • JPE - Identification 2:
    Passes Test
  • JPE - Identification 3:
    Passes Test
  • JPE - Moisture Content:
    ≤ 3.0 %
  • JPE - Acid Value:
    ≤ 4.0
  • JPE - Saponification Value (mg KOH/g):
    43 - 55
  • JPE - pH:
    4.0 - 7.0
  • JPE - Residue on Ignition:
    ≤ 0.25 %
  • JPE - Specific Gravity at 20°C:
    1.090 - 1.130
  • JPE - Viscosity (mm²/S):
    350 - 550
  • JPE - Heavy Metals (as Pb):
    ≤ 20 ppm
  • JPE - Arsenic (As):
    ≤ 2 ppm
  • Ethylene glycol:
    ≤ 0.10 %
  • Diethylene glycol :
    ≤ 0.10 %

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    20 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.