Objectives for PSM-1000 microscopes
Near Infrared Ray (NIR) objective (WD = 20,5 mm)
Catalog # MOTI1101001703531
Supplier: MOTIC
Jump to:
Product Details & Documents
Specifications
- Magnification:20×/0,4
- Description:Near Infrared Ray (NIR) objective (WD = 20,5 mm)
- Seq:111