Order Entry
Italy
Orders LinkContactUsLinkComponent
 

HEPES, 1,0 M buffer solutions

HEPES buffer solution, 1 mol/L, 8,0, Glass bottle, 500 mL
  J63578.AP
HEPES, 1,0 M buffer solutions
  J63578.AP
 :  
HEPES, 1,0 M buffer solutions
HEPES, 1,0 M buffer solutions

HEPES buffer solution, 1 mol/L, 8,0, Glass bottle, 500 mL

 :  
 
210,00 €
   
1 * 500 mL
 

 

  • Concentration:
    1 mol/L
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    HEPES buffer solution
  • pH value:
    8,0
  • Cat. No.:
    J63578.AP
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006158
  • CAS No.:
    7365-45-9

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.