Order Entry
Italy
Orders LinkContactUsLinkComponent
Toremifene, ≥98% 1 * 1 g

Toremifene, ≥98% 1 * 1 g

Catalog # J63803.03
Toremifene, ≥98% 1 * 1 g
Catalog # J63803.03
CAS Number:  
Toremifene, ≥98% 1 * 1 g
CAS Number:  
Price
182,00 €
Log in for availability
1 * 1 g
Product Details & Documents