HEPES, 1,0 M buffer solutions
HEPES buffer solution, 1 mol/L, 8,5, Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 7365-45-9
MDL No.: MFCD00006158
Specifications
Specifications
- Concentration:1 mol/L
- Pack type:Glass bottle
- Pk:250 mL
- Description:HEPES buffer solution
- pH value:8,5



