Succinate, 0,2 M buffer solution, pH 6,5
Succinate buffer solution, 0,2 M, 6,5, 250 mL
Supplier: Thermo Fisher Scientific
Succinate, 0,2 M buffer solution, pH 6,5
Catalog # J62078.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
CAS No.: 150-90-3
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002789
Documents
Specifications
Specifications
- Concentration:0,2 M
- Pk:250 mL
- Description:Succinate buffer solution
- pH value:6,5
- Cat. No.:J62078.AK
- Odour/scent:Odourless
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00002789
- Appearance:Colourless
- CAS No.:150-90-3
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



