Order Entry
Italy
Orders LinkContactUsLinkComponent
HEPES, 1,0 M buffer solutions

HEPES, 1,0 M buffer solutions

HEPES buffer solution, 1 mol/L, 9,0, Glass bottle, 500 mL
Catalog # J60440.AP
HEPES, 1,0 M buffer solutions
Catalog # J60440.AP
CAS Number:  
HEPES, 1,0 M buffer solutions
HEPES, 1,0 M buffer solutions

HEPES buffer solution, 1 mol/L, 9,0, Glass bottle, 500 mL

CAS Number:  
Price
231,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1 mol/L
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    HEPES buffer solution
  • pH value:
    9,0
  • Cat. No.:
    J60440.AP
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006158
  • CAS No.:
    7365-45-9

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.