Buffers, MOPS, 0,5 M solutions
MOPS buffer solution, 0,5 mol/L, 7,0, Glass bottle, 18, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 1132-61-2
Odour: odourless
MDL No.: MFCD00006183
Documents
Specifications
Specifications
- Concentration:0,5 mol/L
- Pack type:Glass bottle
- Pk:100 mL
- Description:MOPS buffer solution
- Pk Info:18
- pH value:7,0
- Cat. No.:J60328.AE
- Odour/scent:Odourless
- Physical form:Liquid
- MDL No.:MFCD00006183
- CAS No.:1132-61-2



