MOPS-EDTA-sodium acetate buffer (MESA)
MOPS-EDTA-sodium acetate buffer (MESA), 1 L
Supplier: Thermo Fisher Scientific
MOPS-EDTA-sodium acetate buffer (MESA)
Catalog # J60184.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
MDL No.: MFCD00131625
Documents
Specifications
Specifications
- Pk:1 L
- Description:MOPS-EDTA-sodium acetate buffer (MESA)
- Cat. No.:J60184.K2
- Physical form:Liquid
- MDL No.:MFCD00131625
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



