Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Lecithin (from soy bean) ≥90%

100 g
Catalog # J61675.22
Lecithin (from soy bean) ≥90%
Catalog # J61675.22
CAS Number:  8002-43-5
Lecithin (from soy bean) ≥90%
Lecithin (from soy bean) ≥90%

100 g

CAS Number:  8002-43-5
Price
50,20 €
Log in for availability
1 * 100 g
Product Details & Documents

Applications: Naturally occuring lipid that protects cells from oxidation. Useful as an emulsifier

  • Pale brown to yellow Solid
  • Odourless
  • Beilstein Reference: 5209585
  • CAS Number:8002-43-5
  • MDL Number:MFCD00147406
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pk:
    100 g

Specifications