Order Entry
Italy
Orders LinkContactUsLinkComponent
HEPES, 1,0 M buffer solutions

HEPES, 1,0 M buffer solutions

HEPES buffer solution, 1 mol/L, 7,5, Glass bottle, 500 mL
Catalog # J60712.AP
HEPES, 1,0 M buffer solutions
Catalog # J60712.AP
CAS Number:  
HEPES, 1,0 M buffer solutions
HEPES, 1,0 M buffer solutions

HEPES buffer solution, 1 mol/L, 7,5, Glass bottle, 500 mL

CAS Number:  
Price
221,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1 mol/L
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    HEPES buffer solution
  • pH value:
    7,5
  • Cat. No.:
    J60712.AP
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006158
  • CAS No.:
    7365-45-9

Specifications