Order Entry
Italy
Orders LinkContactUsLinkComponent
 

CF® Dye cAMP

CF® 640R cAMP, 642/662 nm, 100 µG
Catalog # 00037.
Supplier:   Biotium
CF® Dye cAMP
Catalog # 00037.
Supplier:  Biotium
CAS Number:  
CF® Dye cAMP

CF® 640R cAMP, 642/662 nm, 100 µG

CAS Number:  
Price
204,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Fluorescently labelled cAMP analogs that can be used to probe cAMP receptors.

  • Superior CF® dyes are bright, photostable and water-soluble
  • Available with green or far-red labels
  • Soluble in H₂O, DMF, or DMSO

CF®488A-cAMP and CF®640R-cAMP are fluorescently labelled cAMP analogs that can be used to probe cAMP receptors.

Biotium’s next-generation CF® Dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.

CF is a registered trademark of Biotium, Inc. Alexa Fluor, Texas Red, and DyLight are registered trademarks of Thermo Fisher Scientific.

Specifications

Specifications

  • Pk:
    100 µG
  • Description:
    CF® 640R cAMP
  • Ex/Em:
    642/662 nm

Specifications