Order Entry
Italy
ContactUsLinkComponent
30COMPOSES DANS ACETONE Pyriproxyfen [CAS:95737-68-1] 0.625ug/ml Pirimiphos-methyl [CAS:29232-93-7] 1.25ug/ml Triazophos [CAS:24017-47-8] 1.25ug/ml Azinphos-ethyl [CAS:2642-71-9] 6.25ug/ml Heptenophos [CAS:23560-59-0] 6.25ug/ml Irgarol 105 1 * 5 Ampoul
Codice catalogo 89260.001
Supplier:  VWR Collection
30COMPOSES DANS ACETONE Pyriproxyfen [CAS:95737-68-1] 0.625ug/ml Pirimiphos-methyl [CAS:29232-93-7] 1.25ug/ml Triazophos [CAS:24017-47-8] 1.25ug/ml Azinphos-ethyl [CAS:2642-71-9] 6.25ug/ml Heptenophos [CAS:23560-59-0] 6.25ug/ml Irgarol 105 1 * 5 Ampoul
Codice catalogo 89260.001
Supplier:  VWR Collection
Frequently Bought Together