Mobilealert, notification by sms in case of any error or alarm, requires option h6 for icp 1 * 1 items
MEMMICP110C3
: MEMMERT
Mobilealert, notification by sms in case of any error or alarm, requires option h6 for icp 1 * 1 items
MEMMICP110C3
: MEMMERT
Frequently Bought Together
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection