Phase separating paper, grade 1PS
40 mm × 50 m
Catalog # WHAT2200-654
Jump to:
Discontinued
Product Details & Documents
1PS phase-separating paper is a hydrophobic filter for routine solvent extraction. Within seconds it retains aqueous phases, while solvent phases flow through.
- Impregnated with stabilised silicone to create a hydrophobic filter
- Multiple separations can be processed in parallel
- No special training required
- Can replace a separatory funnel in certain applications
Solvent extraction can be time-consuming and messy when numerous separations are required
Mixed phases are poured directly into the 1PS phase-separating paper. The solvent phase flows through the paper quickly and cleanly, automatically stopping after the process is complete; all of the aqueous phase is retained. Droplets are automatically separated within seconds.
Documents
Certificates
Search by Lot NumberSpecifications
- Seq:09
- Size:40 mm × 50 m