Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Valacyclovir Related Compounds, Reference Standard

Valacyclovir Related Compound D, USP reference standard, Vial, 15 mg
  1707861.
 :   USP
Valacyclovir Related Compounds, Reference Standard
  1707861.
 :  USP
 :  
Valacyclovir Related Compounds, Reference Standard

Valacyclovir Related Compound D, USP reference standard, Vial, 15 mg

 :  
 
1 210,00 €
   
1 * 15 mg
 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

 

 

  • Pack type:
    Vial
  • Pk:
    15 mg
  • Standards:
    USP reference standard
  • Description:
    Valacyclovir Related Compound D
  • Cat. No.:
    1707861.
  • Formula:
    C₁₅H₂₄N₆O₄
  • Description:
    Valacyclovir Related Compound D
  • Synonym(s):
    2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9yl)methoxy]ethyl N-ethyl-L-valinate
  • CAS No.:
    1346747-69-0

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.