Order Entry
Italy
ContactUsLinkComponent
Temperature chart recorder traces on circular paper-charts (125 mm Ø) the temperature of the appliance with weekly recording. The temperature chart recorder is provided with a transparent perspex cover to allow immediate control of the charts and it 1 * 1 ST
Catalog # EVMDZRG10
Supplier:  EVERMED
Temperature chart recorder traces on circular paper-charts (125 mm Ø) the temperature of the appliance with weekly recording. The temperature chart recorder is provided with a transparent perspex cover to allow immediate control of the charts and it 1 * 1 ST
Catalog # EVMDZRG10
Supplier:  EVERMED
Frequently Bought Together