Tween® 40 (Polysorbate)
Glass bottle, 25 g
TCIAT0544-25G
: TCI
: Polyoxyethylene sorbitan monopalmitate
- :9005-66-7
- :C₆₂H₁₂₂O₂₆
- :MFCD00165345
- :? 100 °C (1013 hPa)
- :∼0.1 °C
- :1,05 g/cm³ (20 °C)
- :≥ 149 °C
- :Ambient
- Appearance:Light yellow to Amber to Dark green clear liquid to cloudy liquid
- Water:max. 5.0 %
- Saponification Value:41.0 to 55.0
- Acid Value:max. 4.0
- Pack type:Glass bottle
- Pk:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



