Span® 40
Glass bottle, 500 g
Catalog # TCIAS0061-500G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Sorbitan monopalmitate, Arlacel 40
- CAS Number:26266-57-9
- Formula:C₂₂H₄₂O₆
- MDL Number:MFCD00080946
- Molecular Weight:402.57 g/mol
- Boiling Pt:? 100 °C (1013 hPa)
- Density:1 g/cm³ (20 °C)
- Flash Pt:≥ 149 °C
- Storage Temperature:Ambient
- EINECS:247-568-8
Specifications
Specification Test Results
- Appearance:Light yellow to Brown to Dark green pellet
- Melting point:45.0 to 48.0 deg-C
- Saponification Value:135.0 to 158.0
Specifications
- Pack type:Glass bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



