Span® 40
Glass bottle, 25 g
TCIAS0061-25G
: TCI
: Sorbitan monopalmitate, Arlacel 40
- :26266-57-9
- :C₂₂H₄₂O₆
- :MFCD00080946
- :402.57 g/mol
- :? 100 °C (1013 hPa)
- :1 g/cm³ (20 °C)
- :≥ 149 °C
- :Ambient
- :247-568-8
- Appearance:Light yellow to Brown to Dark green pellet
- Melting point:45.0 to 48.0 deg-C
- Saponification Value:135.0 to 158.0
- Pack type:Glass bottle
- Pk:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



