Span® 20
Glass bottle, 500 g
Catalog # TCIAS0059-500G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Sorbitan monododecanoate, Arlacel 20, Sorbitan monolaurate
- CAS Number:1338-39-2
- Formula:C₁₈H₃₄O₆
- MDL Number:MFCD00005365
- Molecular Weight:346.46 g/mol
- Merck Index:14,08724
- Density:1,05 g/cm³ (20 °C)
- Flash Pt:204°C
- Storage Temperature:Ambient
- EINECS:215-663-3
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Saponification Value:155.0 to 174.0
Specifications
- Pack type:Glass bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



