3,3'-Dihydroxybenzidine ≥99.0% (by titrimetric analysis)
5 g
Catalog # TCIAD2312-5G
Supplier: TCI
3,3'-Dihydroxybenzidine ≥99.0% (by titrimetric analysis)
Catalog # TCIAD2312-5G
Supplier: TCI
CAS Number: 2373-98-0
Jump to:
Product Details & Documents
Synonyms: 4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
- CAS Number:2373-98-0
- Formula:C₁₂H₁₂N₂O₂
- MDL Number:MFCD00039149
- Molecular Weight:216.24 g/mol
Documents
Manual Monomers
Manual
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
- Solubility in N,N-DMF:almost transparency
Specifications
- Pk:5 g



