Order Entry
Italy
Orders LinkContactUsLinkComponent
 

3,3-Dimethylcyclohexanone ≥97.0% (by GC)

Glass bottle, 5 g
Catalog # TCIAD3495-5G
Flammable
Corrosive
Supplier:   TCI
3,3-Dimethylcyclohexanone ≥97.0% (by GC)
Catalog # TCIAD3495-5G
Supplier:  TCI
CAS Number:  2979-19-3
3,3-Dimethylcyclohexanone ≥97.0% (by GC)
3,3-Dimethylcyclohexanone ≥97.0% (by GC)

Glass bottle, 5 g

CAS Number:  2979-19-3
Price
77,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:2979-19-3
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00040178
  • Molecular Weight:126.2 g/mol
  • Boiling Pt:175°C
  • Melting Pt:6 °C
  • Density:0,909 g/cm³ (25 °C)
  • Flash Pt:59°C
  • UN:1224
  • ADR:3,III
  •  :Density: 0.909 g/cm³ (25 °C)
Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications