Order Entry
Italy
Orders LinkContactUsLinkComponent
3,3-Dimethylcyclohexanone ≥97.0% (by GC)

3,3-Dimethylcyclohexanone ≥97.0% (by GC)

Glass bottle, 1 g
Catalog # TCIAD3495-1G
Flammable
Corrosive
Supplier:   TCI
3,3-Dimethylcyclohexanone ≥97.0% (by GC)
Catalog # TCIAD3495-1G
Supplier:  TCI
CAS Number:  2979-19-3
3,3-Dimethylcyclohexanone ≥97.0% (by GC)
3,3-Dimethylcyclohexanone ≥97.0% (by GC)

Glass bottle, 1 g

CAS Number:  2979-19-3
Price
24,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:2979-19-3
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00040178
  • Molecular Weight:126.2 g/mol
  • Boiling Pt:175°C
  • Melting Pt:6 °C
  • Density:0,909 g/cm³ (25 °C)
  • Flash Pt:59°C
  • UN:1224
  • ADR:3,III
  •  :Density: 0.909 g/cm³ (25 °C)
Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.