Order Entry
Italy
Orders LinkContactUsLinkComponent
Digoxin ≥96.0% (by HPLC)

Digoxin ≥96.0% (by HPLC)

Glass bottle, 1 g
Catalog # TCIAD1828-1G
Toxic
Health
Supplier:   TCI
Digoxin ≥96.0% (by HPLC)
Catalog # TCIAD1828-1G
Supplier:  TCI
CAS Number:  20830-75-5
Digoxin ≥96.0% (by HPLC)
Digoxin ≥96.0% (by HPLC)

Glass bottle, 1 g

CAS Number:  20830-75-5
Price
69,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:20830-75-5
  • Formula:C₄₁H₆₄O₁₄
  • MDL Number:MFCD00003674
  • Molecular Weight:780.95 g/mol
  • Merck Index:14,03167
  • Melting Pt:265°C
  • EINECS:244-068-1
  • UN:1544
  • ADR:6.1,II
Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 96.0 area%
  • Specific rotation [a]20/D:
    +10.0 to +13.0 deg (C=10, Pyridine)
  • Solubility in Pyridine:
    almost transparency

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.