Immersion flow cups, ISO 2431
Viscosity immersion flow cup, titanium anodized aluminium, 4 mm
Catalog # 649-0008
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Titanium anodized aluminum or stainless steel cups designed with fixed stainless steel nozzle and handle, for measuring coatings and other fluids.
Certifications: ISO 2431
Documents
TQC Company Video
Videos
Specifications
- Description:Viscosity immersion flow cup, titanium anodized aluminium
- Seq:02
- Cup orifice:4 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Temperature control jacket with tripod for ISO and AFNOR cupsTemperature control jacket with tripod for ISO and AFNOR cupsSIMEVF2068Log in for availability1 * 1 pezzi - 627,00 €
Product Information
- Catalog #AvailabilityPrice
- Digital stopwatch, 0,01 s
Product Information
- Catalog #AvailabilityPrice
- Viscosity convertor of flow time to viscosityViscosity convertor of flow time to viscosity769-1151Log in for availability1 * 1 pezzi - 87,50 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



