Order Entry
Italy
ContactUsLinkComponent
[FR]20 components Methacrifos [CAS:62610-77-9] 20mg/l Phorate [CAS:298-02-2] 20mg/l Phorate-oxon [CAS:2600-69-3] 10mg/l Phorate-sulfone [CAS:2588-04-7] 20mg/l Diazinon [CAS:333-41-5] 20mg/l Disulfoton [CAS:298-04-4] 20mg/l Disulfoton-s [ 1 * 8 Ampoul
Catalog # 89160.001
[FR]20 components Methacrifos [CAS:62610-77-9] 20mg/l Phorate [CAS:298-02-2] 20mg/l Phorate-oxon [CAS:2600-69-3] 10mg/l Phorate-sulfone [CAS:2588-04-7] 20mg/l Diazinon [CAS:333-41-5] 20mg/l Disulfoton [CAS:298-04-4] 20mg/l Disulfoton-s [ 1 * 8 Ampoul
Catalog # 89160.001
Frequently Bought Together