Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories
Accessories
Multi-purpose adapter, MSAA502, for eliminating ambient light or providing an enhanced field of view at optimal distances, compatible with Dino-Lite AM models (excluding models with polariser)
Catalog # IDCPMSAA502
Accessories
Catalog # IDCPMSAA502
Accessories
Accessories
Multi-purpose adapter, MSAA502, for eliminating ambient light or providing an enhanced field of view at optimal distances, compatible with Dino-Lite AM models (excluding models with polariser)
Price
24,20 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Description:
    Multi-purpose adapter, MSAA502, for eliminating ambient light or providing an enhanced field of view at optimal distances, compatible with Dino-Lite AM models (excluding models with polariser)