Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR® Redox Electrodes

VWR® Redox Electrodes

RD 113 (redox), BNC, 1 m cable, Gel, Epoxy, 12×120 mm
Codice catalogo 662-1408
Nome del fornitore:   VWR Collection
VWR® Redox Electrodes
Codice catalogo 662-1408
Nome del fornitore:  VWR Collection
VWR® Redox Electrodes
VWR® Redox Electrodes

RD 113 (redox), BNC, 1 m cable, Gel, Epoxy, 12×120 mm

Price
130,00 €
Log in for availability
1 * 1 pezzi
Specifications
  • Connection:
    BNC, 1 m cable
  • Ø×L:
    12×120 mm
  • Description:
    RD 113 (redox)
  • Material:
    Epoxy
  • Electrolyte:
    Gel