Order Entry
Italy
ContactUsLinkComponent
PRODUCT FOR RNA RELATE APPLICATION, DSRNA MARKER EASY, THE DSRNA MARKER EASY IS SUPPLIED IN A READY-TO-LOAD MIXTURE OF LOADING DYE ( CONTAINING TRIS-HCL BUFFER, GLYCEROL, EDTA SODIUM SALT, SODIUM CHLORIDE, BROMPHENOL BLUE) AND AN IDEAL SIZE MARKER FO 1 * 125 µl
Codice catalogo ABNOR0006
Supplier:  Abnova
PRODUCT FOR RNA RELATE APPLICATION, DSRNA MARKER EASY, THE DSRNA MARKER EASY IS SUPPLIED IN A READY-TO-LOAD MIXTURE OF LOADING DYE ( CONTAINING TRIS-HCL BUFFER, GLYCEROL, EDTA SODIUM SALT, SODIUM CHLORIDE, BROMPHENOL BLUE) AND AN IDEAL SIZE MARKER FO 1 * 125 µl
Codice catalogo ABNOR0006
Supplier:  Abnova
Frequently Bought Together