Order Entry
Italy
ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, H13N8 (A/BLACK-HEADED GULL/NETHERLANDS/1/00) RECOMBINANT PROTEIN, H13N8 (A/BLACK-HEADED GULL/NETHERLANDS /1/00, AAV91212, 17 A.A. - 530 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
Catalog # ABNOP4886
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, H13N8 (A/BLACK-HEADED GULL/NETHERLANDS/1/00) RECOMBINANT PROTEIN, H13N8 (A/BLACK-HEADED GULL/NETHERLANDS /1/00, AAV91212, 17 A.A. - 530 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
Catalog # ABNOP4886
Supplier:  Abnova