Order Entry
Italy
ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, PAK1 (HUMAN) RECOMBINANT PROTEIN, HUMAN PAK1 KINASE DOMAIN (Q13153, 248 A.A. - 545 A.A.) PARTIAL RECOMBI NANT PROTEIN EXPRESSED IN ESCHERICHIA COLI. THE RECOMBINANT PROTEIN DOES NOT HAVE THE INHIBITORY SWITCH DOMAIN AND HA 1 * 10 µG
Catalog # ABNOP5382
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, PAK1 (HUMAN) RECOMBINANT PROTEIN, HUMAN PAK1 KINASE DOMAIN (Q13153, 248 A.A. - 545 A.A.) PARTIAL RECOMBI NANT PROTEIN EXPRESSED IN ESCHERICHIA COLI. THE RECOMBINANT PROTEIN DOES NOT HAVE THE INHIBITORY SWITCH DOMAIN AND HA 1 * 10 µG
Catalog # ABNOP5382
Supplier:  Abnova