Order Entry
Italy
ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, 10 X TAQ BUFFER, 10 X TAQ BUFFER IS A READY-TO-USE BUFFER TO BE USED FOR TAQ, HOT TAQ, RED TAQ A ND LONG TAQ ENZYMES IN PCR REACTIONS. DOES NOT CONTAIN MGCL2. 1 * 1,8 mL
Catalog # ABNOD0024
Supplier:  Abnova
PRODUCT FOR DNA RELATE APPLICATION, 10 X TAQ BUFFER, 10 X TAQ BUFFER IS A READY-TO-USE BUFFER TO BE USED FOR TAQ, HOT TAQ, RED TAQ A ND LONG TAQ ENZYMES IN PCR REACTIONS. DOES NOT CONTAIN MGCL2. 1 * 1,8 mL
Catalog # ABNOD0024
Supplier:  Abnova
Frequently Bought Together