Order Entry
Italy
Orders LinkContactUsLinkComponent
(10S,17S)-DiHDoHE

(10S,17S)-DiHDoHE

25 µG
Catalog # CAYM10008128-25
Supplier:   Cayman Chemical
(10S,17S)-DiHDoHE
Catalog # CAYM10008128-25
Supplier:  Cayman Chemical
CAS Number:  871826-47-0
(10S,17S)-DiHDoHE

25 µG

CAS Number:  871826-47-0
Price
309,00 €
Log in for availability
1 * 25 µG
Product Details & Documents
  • CAS Number:871826-47-0
  • Formula:C₂₂H₃₂O₄
Specifications

Specifications

  • Pk:
    25 µG

Specifications