Order Entry
Italy
Orders LinkContactUsLinkComponent
Estriol

Estriol

Glass bottle for solids, 5 g
Catalog # CAYM10006484-5
Supplier:   Cayman Chemical
Estriol
Catalog # CAYM10006484-5
Supplier:  Cayman Chemical
CAS Number:  50-27-1
Estriol

Glass bottle for solids, 5 g

CAS Number:  50-27-1
Price
650,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:50-27-1
  • Formula:C₁₈H₂₄O₃
  • MDL Number:MFCD00003691
  • EINECS:200-022-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.