Accessories for DOSE IT
Aspiration tube, stainless steel, one end dented, length: 35 cm, 8 mm
Catalog # 710-1040
Supplier: INTEGRA Biosciences
Jump to:
Product Details & Documents
Documents
Dose It
Videos
Specifications
- Description:Aspiration tube, stainless steel, one end dented, length: 35 cm
- Ø int.:8 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



