Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories for DOSE IT
Accessories for DOSE IT
Aspiration, dispensing tube, stainless steel, one end dented, length: 35 cm, 4 mm
Catalog # 710-1038
Supplier:   INTEGRA Biosciences
Accessories for DOSE IT
Catalog # 710-1038
Accessories for DOSE IT
Accessories for DOSE IT
Aspiration, dispensing tube, stainless steel, one end dented, length: 35 cm, 4 mm
Price
50,40 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Documents
Specifications
  • Description:
    Aspiration, dispensing tube, stainless steel, one end dented, length: 35 cm
  • Ø int.:
    4 mm