Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories for DOSE IT
Accessories for DOSE IT
Aspiration, dispensing tube, stainless steel, one end dented, length: 10 cm, 8 mm
Catalog # 710-1037
Supplier:   INTEGRA Biosciences
Accessories for DOSE IT
Catalog # 710-1037
Accessories for DOSE IT
Accessories for DOSE IT
Aspiration, dispensing tube, stainless steel, one end dented, length: 10 cm, 8 mm
Price
29,20 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Documents
Specifications
  • Description:
    Aspiration, dispensing tube, stainless steel, one end dented, length: 10 cm
  • Ø int.:
    8 mm