Veterinary needles
Veterinary Luer needles, 12, 25 mm
Catalog # POPR7230
Supplier: CADENCE SCIENCE
Jump to:
Product Details & Documents
These hypodermic needles are all fitted with a large knurled nickel plated hub which allows for secure gripping even under the most difficult circumstances.
Specifications
- Gauge:12
- Description:Veterinary Luer needles
- Length:25 mm
- Seq:02
- Cat. No.:POPR7230