Tryptone Base Materials
Tryptone, Glass bottle for solids, 250 g
Catalog # RC-111
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Tryptone is an assortment of peptides generated by the tryptic digestion of casein. It is commonly used in microbiology to produce Lysogeny broth for the growth of E. coli and other microorganisms, as it provides a source of amino acids.
Available in different pack sizes
Solubility: Water
- Appearance: Cream to beige free flowing powder
- CAS: 91079-40-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Description:Tryptone
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



