Order Entry
Italy
Orders LinkContactUsLinkComponent
Tryptone Base Materials

Tryptone Base Materials

Tryptone, Glass bottle for solids, 500 g
Catalog # RC-110
Supplier:   G-Biosciences
Tryptone Base Materials
Catalog # RC-110
Supplier:  G-Biosciences
CAS Number:  
Tryptone Base Materials

Tryptone, Glass bottle for solids, 500 g

CAS Number:  
Price
172,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Tryptone is an assortment of peptides generated by the tryptic digestion of casein. It is commonly used in microbiology to produce Lysogeny broth for the growth of E. coli and other microorganisms, as it provides a source of amino acids.

  • Available in different pack sizes

    Solubility: Water

  • Appearance: Cream to beige free flowing powder
  • CAS: 91079-40-2

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Description:
    Tryptone

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.