Tryptone Base Materials
Tryptone, Bottiglia di vetro, 100 g
Catalog # RC-109
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Tryptone is an assortment of peptides generated by the tryptic digestion of casein. It is commonly used in microbiology to produce Lysogeny broth for the growth of E. coli and other microorganisms, as it provides a source of amino acids.
Available in different pack sizes
Solubility: Water
- Appearance: Cream to beige free flowing powder
- CAS: 91079-40-2
Specifications
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:100 g
- Descrizione:Tryptone



