Order Entry
Italy
Orders LinkContactUsLinkComponent
Stirrer Guides with Anti-clockwise Thread
Stirrer Guides with Anti-clockwise Thread
Stirrer guide, dynamic double seal, 29/32, 10 mm
Catalog # LENZ06306004
Supplier:   Lenz Laborglas
Stirrer Guides with Anti-clockwise Thread
Catalog # LENZ06306004
Supplier:  Lenz Laborglas
Stirrer Guides with Anti-clockwise Thread
Stirrer Guides with Anti-clockwise Thread
Stirrer guide, dynamic double seal, 29/32, 10 mm
Price
328,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
DURAN®, borosilicate glass, PFTE/fibre glass pressure screw. The fluoroplastic bearing is in a borosilicate glass housing, and has adjustable PTFE/FPM seal. All parts are interchangeable.
Specifications
  • NS:
    29/32
  • Description:
    Stirrer guide, dynamic double seal
  • Seq:
    02
  • Shaft Ø:
    10 mm