Order Entry
Italy
ContactUsLinkComponent
KIT, TNFRSF11B (HUMAN) ELISA KIT, TNFRSF11B (HUMAN) ELISA KIT IS A BIOTIN LABELLED ANTIBODY BASED SANDWICH ENZYME IMMUNOASSAY FOR TH E QUANTITATIVE MEASUREMENT OF HUMAN OSTEOPROTEGERIN IN SERUM, PLASMA, SYNOVIAL FLUID AND TISSUE CULTURE MEDIUM. 1 * 1 KIT
Catalog # ABNOKA0030
Supplier:  Abnova
KIT, TNFRSF11B (HUMAN) ELISA KIT, TNFRSF11B (HUMAN) ELISA KIT IS A BIOTIN LABELLED ANTIBODY BASED SANDWICH ENZYME IMMUNOASSAY FOR TH E QUANTITATIVE MEASUREMENT OF HUMAN OSTEOPROTEGERIN IN SERUM, PLASMA, SYNOVIAL FLUID AND TISSUE CULTURE MEDIUM. 1 * 1 KIT
Catalog # ABNOKA0030
Supplier:  Abnova