Order Entry
Italy
Orders LinkContactUsLinkComponent
 

VWR®, Test Tubes Racks

Orange, 24 (3×8), 30 mm, 110×285,5×86,7 mm
Catalog # 212-0427
Supplier:   VWR Collection
VWR®, Test Tubes Racks
Catalog # 212-0427
Supplier:  VWR Collection
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Orange, 24 (3×8), 30 mm, 110×285,5×86,7 mm

Price
31,50 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Specifications
  • For tube Ø:
    30 mm
  • No. of Holes:
    24 (3×8)
  • Colour:
    Orange
  • Seq:
    15
  • W×D×H:
    110×285,5×86,7 mm