Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Test Tubes Racks

VWR®, Test Tubes Racks

Blue, 20 (4×5), 20 mm, 130×105×84 mm
Catalog # 212-0390
Supplier:   VWR Collection
VWR®, Test Tubes Racks
Catalog # 212-0390
Supplier:  VWR Collection
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Blue, 20 (4×5), 20 mm, 130×105×84 mm

Price
21,20 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Specifications
  • For tube Ø:
    20 mm
  • No. of Holes:
    20 (4×5)
  • Colour:
    Blue
  • Seq:
    07
  • W×D×H:
    130×105×84 mm