Order Entry
Italy
Orders LinkContactUsLinkComponent
 

VWR®, Test Tubes Racks

Green, 20 (4×5), 20 mm, 130×105×84 mm
Codice catalogo 212-0391
Nome del fornitore:   VWR Collection
VWR®, Test Tubes Racks
Codice catalogo 212-0391
Nome del fornitore:  VWR Collection
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Green, 20 (4×5), 20 mm, 130×105×84 mm

Price
21,20 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Specifications
  • For tube Ø:
    20 mm
  • No. of Holes:
    20 (4×5)
  • Colour:
    Green
  • Seq:
    08
  • W×D×H:
    130×105×84 mm