Order Entry
Italy
Orders LinkContactUsLinkComponent
 

VWR®, Test Tubes Racks

Orange, 72 (6×12), 13 mm, 103,5×202,5×60 mm
  212-0403
VWR®, Test Tubes Racks
  212-0403
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks

Orange, 72 (6×12), 13 mm, 103,5×202,5×60 mm

 
23,80 €
   
1 * 1 pezzi
 

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
 
  • For tube Ø:
    13 mm
  • No. of Holes:
    72 (6×12)
  • Colour:
    Orange
  • Seq:
    51
  • W×D×H:
    103,5×202,5×60 mm